Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chromogenic Salmonella Agar Base (Dehydrated)

Catalog No. OXCM1007B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM1007B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM1007B Supplier Thermo Scientific™ Supplier No. CM1007B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and differentiate Salmonella species from other Enterobacteriaceae with this selective medium.

Isolate and differentiate Salmonella species from other Enterobacteriaceae with Thermo Scientific™ Oxoid™ Salmonella Chromogenic Agar Base (Dehydrated), designed to identify Salmonella species based on their utilization of one chromogenic substrate. Their inability to utilize another chromogenic substrate that most other members of the family Enterobacteriaceae can utilize enables rapid and reliable identification of Salmonella species.

Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.

Isolate and differentiate Salmonella species from other Enterobacteriaceae with Salmonella Chromogenic Medium.

  • Easy to read: a combination of two chromogens visually differentiates Salmonella from other Enterobacteriaceae.
  • Selective for Salmonella: due to the additions of bile salts, novobiocin, and cefsulodin.

Traditionally, media used to differentiate Salmonella species from other members of the family Enterobacteriaceae depend upon the ability of Salmonella species to produce hydrogen sulphide coupled with their ability to ferment lactose.1, 2 These are, however, essentially inadequate methods with a significant number of species not exhibiting these characteristics. Chromogenic media have been developed for rapid and more reliable identification of Salmonella.

Salmonella Chromogenic Medium combines two chromogens for the detection of Salmonella species, 5-Bromo-6-Chloro-3-Indolyl caprylate (Magenta-caprylate) and 5-Bromo-4-Chloro-3-Indolyl b-D galactopyranoside (X-gal). X-gal is a substrate for the enzyme b-D-galactosidase. Most Enterobacteriaceae are b-D-galactosidase positive and produce blue colored colonies on Salmonella Chromogenic Medium. Hydrolysis of the chromogen Mag-caprylate by lactose negative Salmonella species results in magenta colonies which are easily distinguished from the blue colored colonies of other organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Salmonella Chromogenic Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.0L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.