Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Columbia Blood Agar Base (Dehydrated)

Catalog No. p-4524643
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate fastidious microorganisms with clear hemolytic reactions and good colonial differentiation with this general-purpose blood agar base.

Cultivate and isolate fastidious microorganisms with clearly visible hemolytic reactions with Thermo Scientific™ Oxoid™ Columbia Blood Agar Base (Dehydrated) when used with whole defibrinated blood (SR0050). Columbia Blood Agar Base is an ideal base medium for preparation of blood agar, chocolate agar and for preparation of various selective and identification media.

Grow fastidious organisms with rapid production of large colonies, clearly defined zones of hemolysis and good colonial differentiation for all-around performance with Columbia Blood Agar when used with whole defibrinated blood.

Traditionally, blood agar has been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and the second in clearly defined zones of hemolysis and good colonial differentiation. This formulation combines the virtues of both for optimal performance1.

  • Produces large colonies of fastidious microorganisms with clearly defined hemolytic reactions and colonial morphology.
  • Serves as an ideal base for the media containing blood and for selective media formulations in which different combinations of antimicrobial agents are used as additives.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Columbia Blood Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.8L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.