Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Oxoid™ Cronobacter Screening Broth Base

Catalog No. cm1121b
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1121B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CM1121B Supplier Thermo Scientific™ Supplier No. CM1121B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Oxoid Cronobacter Screening Broth Base, a selective enrichment broth, isolates Cronobacter spp, from food and environmental samples.

Oxoid Cronobacter Screening Broth Base (CSB) is a selective enrichment broth for isolation of Cronobacter spp, from food and environmental samples.

Requires Vancomycin Supplement, 5mg, Part No. SR0247E.
TRUSTED_SUSTAINABILITY

Specifications

Description Cronobacter Screening Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 17.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.