Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dermasel Agar Base (Dehydrated)

Catalog No. cm0539b
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and identify dermatophyte fungi from hair, nails or skin scrapings using this selective medium.

Isolate and identify dermatophyte fungi from hair, nails or skin scrapings using Thermo Scientific™ Oxoid™ Dermasel Agar Base (Dehydrated) when used with Dermasel Selective Supplement (SR0075). The near neutral pH of the medium is better for the growth of fungi. The often-recommended acidic pH used to supress bacterial contaminants is replaced by addition of selective agents. Chloramphenicol and cycloheximide render the medium selective for dermatophytes inhibiting many saprophytic fungi, yeasts and bacterial skin flora1.

Use Dermasel Agar Base for the isolation of dermatophyte fungi from samples such as hair, skin scrapings and nails.

  • Inhibits the growth of contaminating saprophytic fungi, yeasts and bacterial skin flora1.
  • Chloramphenicol and cycloheximide in the medium improve the isolation of dermatophytes, especially when the inoculum is heavily contaminated2, 3

The chloramphenicol and cycloheximide supplement SR0075 reduces the potential risk to health from these antibiotics. To include them in the powder mix could allow them to be scattered as dust while weighing the medium. It also ensures a fixed, accurate dose of antibiotic that has been protected from degradation on storage.

The addition of cycloheximide and an anti-bacterial agent has been reported to improve considerably the isolation of dermatophytes, especially when the inoculum, such as horse hair, was heavily contaminated3,4. The presence of staphylococci, which may grow in the absence of the antibiotic has been shown to prevent the in-vitro growth of Trichophyton rubrum5.

The presence of cycloheximide in the medium inhibits the growth of Trichosporon cutaneum, Candida parasilosis, Candida krusei, Aspergillus, Penicillium, Fusarium and Cephalosporium species which have been associated with diseased nails.6,7

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Dermasel Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.2L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.