Learn More
Thermo Scientific™ M17 Agar (Dehydrated)
Description
Cultivate lactic streptococci and their bacteriophages and selectively enumerate Streptococcus thermophiles from yogurt with this medium.
Cultivate lactic streptococci and their bacteriophages and selectively enumerate Streptococcus thermophilus from yogurt with Thermo Scientific™ Oxoid™ M17 Agar (Dehydrated). M17 Agar is based on the formulation described by Terzaghi and Sandine1 and is recommended as an improved medium for the growth and enumeration of lactic streptococci and their bacteriophages.
Lactic streptococci are nutritionally fastidious and require complex media for optimal growth2,3. Their homofermentative acid-producing nature require that the medium is well buffered so that the culture pH is maintained above 5.7 during active growth. This maintenance of the pH is important as lower pH can results in injury and reduced recovery of lactic streptococci.
M17 Agar contains di-sodium glycerophosphate which has sufficient buffering capacity to maintain the pH above 5.7 of actively growing cultures even after 24 hours at 30°C. This buffering agent also allows the addition of calcium without a precipitation complex being formed. The calcium-containing medium is used for the assay of bacteriophages of lactic streptococci1
Shankar and Davies4 reported that M17 Agar was suitable for the isolation and enumeration of Streptococcus thermophilus from yogurt as the high concentration of di-sodium-glycerophosphate resulted in suppression of Lactobacillus bulgaricus. M17 Agar has been recommended5,6 by the International Dairy Federation for the selective enumeration of Streptococcus thermophilus from yogurt.
M17 Agar is also suitable for growing and maintaining starter cultures for cheese and yogurt manufacture as it has little deleterious effect on their subsequent acid-producing ability in milk at either 30°C or 22°C 1.
One further useful property of this agar is its ability to detect streptococcal mutants which are unable to ferment lactose1. These mutant Lac- strains form much smaller colonies than the parent lactose fermenting strain.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Specifications
Certifications/Compliance | Industrial Reference: IDF. |
Description | M17 Agar |
Form | Powder |
Product Type | Agar |
Quantity | 500 g |
Yield | For 10.4L medium |
For Laboratory Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.