Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MacConkey Agar without Salt (Dehydrated)

Catalog No. p-4525021
Click to view available options
Quantity:
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Detect and enumerate coliform organisms in clinical samples with this medium.

Detect and enumerate coliform organisms in clinical samples with Thermo Scientific™ Oxoid™ MacConkey Agar without Salt (Dehydrated). The differential medium suppresses swarming of Proteus species and is, therefore, recommended for urine examination in the investigation of suspected urinary tract infection (UTI).

MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.

TRUSTED_SUSTAINABILITY

Specifications

Description MacConkey Agar w/o Salt (Dehydrated)
Form Powder
Product Type Agar
Quantity 2.5 kg
Yield for 53.2L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.