Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Modified Semi-Solid Rappaport-Vassiliadis (MRSV) Medium Base (Dehydrated)

Catalog No. oxcm0910b
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
OXCM0910B 500 g
1 options
This item is not returnable. View return policy

Catalog No. OXCM0910B

Supplier: Thermo Scientific™ CM0910B

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect motile Salmonella species from food and environmental samples with this semi-solid, selective medium.

Detect motile Salmonella species from food and environmental samples with Thermo Scientific™ Oxoid™ Modified Semi-Solid Rappaport-Vassiliadis Medium Base (Dehydrated). Motility enrichment on MSRV Medium has been designed as a simple, sensitive method for the isolation of salmonellae. The efficacy of the medium is based upon the ability of salmonellae to migrate through the selective medium ahead of competing Gram-negative, motile organisms and, therefore, produce opaque halos of growth.

Utilize motility enrichment on Modified Semi-solid Rappaport Vassiliadis Agar for the isolation of Salmonella species from food and environmental samples associated with the primary stage of animal production (such as dust).

  • A simple and sensitive method for the isolation of salmonellae from food and environmental samples.
  • Gram-positive flora is inhibited by novobiocin, low pH, high magnesium chloride concentration, and malachite green.
  • Isolation is based on the ability of salmonellae to migrate through the selective medium ahead of competing Gram-negative, motile organisms, thus producing opaque halos of growth.

Modified Semi-solid Rappaport Vassiliadis (MSRV) Medium is based on the formulation described by De Smedt et al. which has been shown to detect more Salmonella-positive samples than the traditional enrichment procedures1,2. Further collaborative studies have confirmed these findings3,4.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL.
Description Modified Semi-Solid Rappaport-Vassiliadis Medium Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 15.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.