Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Mycoplasma Broth Base (Dehydrated)

Catalog No. oxcm0403b
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Catalog No. OXCM0403B

Supplier: Thermo Scientific™ CM0403B

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and cultivate mycoplasmas from clinical specimens with this medium.

Isolate and cultivate mycoplasmas from clinical specimens with Thermo Scientific™ Oxoid™ Mycoplasma Broth Base (Dehydrated) when supplemented with Supplement- G Selective Supplement SR0059. Mycoplasma Broth Base complements Mycoplasma Agar Base CM0401.

Mycoplasma are a mollicute genus of bacteria that lack a cell wall around their cell membranes. This characteristic makes them naturally resistant to many common antibiotics such as penicillin or other beta-lactam antibiotics that target cell wall synthesis. They can be parasitic or saprotrophic. They are mainly parasites on the surface of human and animal epithelium cells.

Mycoplasma Broth Base is formulated as a basal medium to be enriched with any satisfactory supplementary factors used for the growth of mycoplasmas.

Most strains of mycoplasmas are encouraged by growth in a biphasic medium in which a layer of Mycoplasma Broth Base covers a basal layer of Mycoplasma Agar Base CM0401.

The majority of mycoplasma require media enriched with serum for good growth. Also antibacterial agents are necessary to prevent overgrowth of the slow-growing mycoplasmas by contaminating organisms.

Mycoplasma Supplement-G SR0059 is a general supplement that when added to Mycoplasma Broth Base provides the necessary factors for growth and prevents overgrowth of slow-growing contaminating organisms.   

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Mycoplasma Broth Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 19.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.