Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Agar (Dehydrated)

Catalog No. OXCM0003B
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Catalog No. OXCM0003B

Supplier: Thermo Scientific™ CM0003B

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate and isolate fastidious pathogens and other micro-organisms with this nutritive, non-selective medium.

Cultivate and isolate fastidious pathogens and other microorganisms with Thermo Scientific™ Oxoid™ Nutrient Agar (Dehydrated). This non-selective medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

Nutrient Agar is a basic culture medium used to subculture organisms for maintenance purposes or to check the purity of subcultures from isolation plates prior to biochemical or serological tests.

In semi-solid form, as agar slopes or agar butts, the medium is used to maintain control organisms1.

Nutrient Agar is suitable for teaching and demonstration purposes. It contains a concentration of 1.5% agar to permit the addition of up to 10% blood or other biological fluid, as required. The medium, without additions, may be used for the cultivation of organisms which are not exacting in their nutritional requirements.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, FDA, ISO, NMKL.
Description Nutrient Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 17.9L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.