Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Rose-Bengal Chloramphenicol Agar Base (Dehydrated)

Catalog No. OXCM0549B
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and enumerate yeasts and molds from a wide variety of foodstuffs with this selective medium.

Isolate and enumerate yeasts and molds from a wide variety of foodstuffs with Thermo Scientific™ Oxoid™ Rose Bengal Chloramphenicol Agar Base (Dehydrated). The neutral pH of the medium and the presence of selective agent chloramphenicol helps to suppress the bacterial growth, allowing molds and yeasts to grow. Rose Bengal in the medium is taken up by mold and yeast colonies assisting the enumeration of small colonies1. Rose Bengal also controls the size and height of mold colonies preventing the overgrowth, resulting in improved enumeration and detection.

Use Rose Bengal Chloramphenicol Agar for selective enumeration of molds and yeast from foods

  • Easy colony acounting: Rose-Bengal in the medium controls the size and height of mold colonies preventing overgrowth and making colony counting easier.
  • Selective: appropriate for isolation of yeasts and molds from foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria
  • Suitable for higher and prolonged incubation temperatures.

The choice of a suitable medium for enumeration of yeasts and molds is greatly dependent on the nature of the foodstuffs under investigation and the organisms that occur on them2. Rose-Bengal Chloramphenicol Agar is recommended for fresh proteinaceous foods whose associated flora consists mainly of Gram-negative rod-shaped bacteria, although it should be noted that chloramphenicol alone may not be sufficient to inhibit the bacterial background. Because of the stability of chloramphenicol, Rose-Bengal Chloramphenicol Agar is also suitable when higher and prolonged incubation temperatures around 35°C are required.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Rose-Bengal Chloramphenicol Agar Base
Form Powder
Product Type Agar
Quantity 500 g
Yield For 15.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.