Learn More
Thermo Scientific™ Sorbitol MacConkey Agar (Dehydrated)
Description
Selectively detect Escherichia coli O157 with this medium.
Selectively detect Escherichia coli O157 with Thermo Scientific™ Oxoid™ Sorbitol MacConkey Agar (Dehydrated). The formulation, based on that described by Rappaport and Henig1 is identical to MacConkey Agar No.3, except that lactose has been replaced with sorbitol. Escherichia coli O157 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E. coli strains ferment sorbitol and form pink colonies.
Escherichia coli O157 is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhea and severe abdominal pain, and hemolytic uraemic syndrome (HUS) 2, 3, 4, 5,6, and as such, is a significant human pathogen.
Sorbitol MacConkey Agar is recommended for the isolation of pathogenic E.coli O157.
- Sensitive and specific for coli O157 based on sorbitol fermentation.
- Recommended as a simple, inexpensive, rapid and reliable means of screening for E.coli O157
The efficiency of Sorbitol MacConkey Agar has been confirmed by March and Ratnam7. These workers reported a sensitivity of 100% and a specificity of 85% and recommended the medium as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications
Specifications
Certifications/Compliance | Industrial Reference: FDA. |
Description | Sorbitol MacConkey Agar |
Form | Powder |
Product Type | Agar |
Quantity | 500 g |
Yield | For 9.7L medium |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.