Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptone Bile X-Glucuronide (TBX) Medium (Dehydrated)

Catalog No. r669452
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Detect and enumerate Escherichia coli in food and animal feed samples with this chromogenic medium while adhering to ISO 11133:2014.

Detect and enumerate Escherichia coli in food and animal feed samples with Thermo Scientific™ Oxoid™ Typtone Bile X-Glucuronide (TBX) Medium (Dehydrated). The medium meets the formulation and performance criterion laid down in ISO 166491, 2, 3 and ISO 11133:20144.

TBX Medium builds on the speed, reliability and recovery from frozen samples and detection of poor lactose fermenters offered by the original Tryptone Bile Agar4,5, with the addition of a chromogenic agent, X-glucuronide, which detects glucuronidase activity, an enzyme shown to be highly specific for Escherichia coli6.

Use TBX Medium for selective detection and enumeration of Escherichia coli in food and animal feed samples.

  • Selective: presence of bile salts in the medium provide selectivity against Gram-positive bacteria, particularly bacilli and fecal streptococci.
  • Easy-to-read: due to the addition of chromogenic 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide (X-glucuronide), E. coli appear as blue/green colonies.
  • ISO compliant: meets the formulation and performance criteria laid down in ISO 16649 and ISO 11133:2014.

Most Escherichia coli strains can be differentiated from other coliforms by the presence of the enzyme glucuronidase. However, approximately 3 to 4% of Escherichia coli are glucuronidase negative, notably Escherichia coli O157 strains7.

The chromogen in TBX Medium is 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide (X-glucuronide) and is targeted by glucuronidase. Escherichia coli cells are able to absorb this complex intact and intracellular glucuronidase splits the bond between the chromophore and the glucuronide. The released chromophore is colored and builds up within the cells, causing Escherichia coli colonies to be colored blue/green.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description TBX Medium (Dehydrated)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 13.7L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.