Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Urea Broth Base (Dehydrated)

Catalog No. OXCM0071B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0071B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0071B Supplier Thermo Scientific™ Supplier No. CM0071B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate urease- producing Enterobacteriaceae from members of the Salmonella and Shigella groups with this medium.

Differentiate urease-producing Enterobacteriaceae from members of the Salmonella and Shigella groups, during the routine examination of rectal swabs and feces with Thermo Scientific™ Oxoid™ Urea Broth Base (Dehydrated). This is a liquid modification of Christensen medium1introduced by Maslen.

This modification allows inoculation by Pasteur pipette, and it is easier to detect contamination during storage. Maslen also claimed that it is easier to detect positive results.

Maslen noted that in the routine examination of feces for Salmonella and Shigella organisms, many non-lactose-fermenting colonies isolated were later found to belong to the urease-positive Proteae. He evolved this medium as a means whereby the latter organisms could be rapidly detected and eliminated - thus saving a considerable amount of time and media. Maslen claimed that the advantages of the fluid medium were:
1. A Pasteur pipette could be used to inoculate other diagnostic media.
2. Rapid growth ensued and it was possible to discern a clear-cut positive reaction within two to five hours at 35°C.
3. It was easier to detect any contamination during storage.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, FDA.
Description Urea Broth Base (Christensen Broth Base)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 55.6L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.