Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WL Nutrient Broth (Dehydrated)

Catalog No. cm0501b
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Determine the flora in brewing and fermentation processes with this selective medium.

Determine the flora in brewing and fermentation processes with Thermo Scientific™ Oxoid™ WL Nutrient Broth (Dehydrated). The broth is based on the formulation of Green and Gray1 and is used where there are advantages for broth media e.g. using larger samples of liquid products or for enrichment cultures with cycloheximide.

Use WL Nutrient Broth for determination of the flora in brewing and fermentation processes.

  • Selective: the presence of cycloheximide in the medium suppresses the yeast growth and renders the medium selective for bacterial contaminants
  • Baker’s yeast counts can be carried out in this medium at a pH 5.5, while at pH 6.5, the medium can be used for obtaining counts of Baker’s and distiller’s yeast.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Description WL Nutrient Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 8.3L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.