Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXPAT Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1106011A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106011A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106011A32 Supplier Novus Biologicals Supplier No. NB11060511AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OXPAT Polyclonal specifically detects OXPAT in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen OXPAT
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5
Gene Symbols PLIN5
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse OXPAT protein, within residues 400-463. [UniProtNo. Q8BVZ1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 440503
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.