Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXPAT Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11060509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060509 0.1 mL
NB11060509T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11060509 Supplier Novus Biologicals Supplier No. NB11060509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

OXPAT Polyclonal specifically detects OXPAT in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen OXPAT
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2.0 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1.0-5.0 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26096040), Immunohistochemistry-Paraffin 1.0-5.0 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 24648545)
Formulation PBS and 30% Glycerol with 0.1% Sodium Azide
Gene Accession No. Q00G26
Gene Alias lipid storage droplet protein 5, LSDA5, LSDP5, MLDP, OXPAT, perilipin 5, perilipin-5
Gene Symbols PLIN5
Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human OXPAT protein, within residues 300-400. [UniProt# Q00G26]
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Lipid Droplets
Primary or Secondary Primary
Gene ID (Entrez) 440503
Test Specificity Human, bovine and mouse. Immunogen has 94% homology to sheep and porcine and 84% homology to rat.
Target Species Human, Mouse, Pig, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.