Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXSM Antibody (OTI4E10), PerCP, Novus Biologicals™
SDP

Catalog No. NB302148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302148 Supplier Novus Biologicals Supplier No. NBP273176PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OXSM Monoclonal antibody specifically detects OXSM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen OXSM
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4E10
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias 3-ketoacyl-acyl carrier protein synthase, 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial, 3-oxoacyl-ACP synthase, mitochondrial, acyl-malonyl acyl carrier protein-condensing enzyme, beta-ketoacyl synthase, Beta-ketoacyl-ACP synthase, EC 2.3.1, EC 2.3.1.41, FASN2D, FLJ20604, KASI, KS3-oxoacyl-acyl-carrier-protein synthase, type II mitochondrial beta-ketoacyl synthase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 78-343 of human OXSM (NP_060367) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54995
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.