Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxyntomodulin Antibody (20), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB163356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163356 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163356 Supplier Novus Biologicals Supplier No. NBP261945JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Oxyntomodulin Monoclonal antibody specifically detects Oxyntomodulin in Human samples. It is validated for ELISA

Specifications

Antigen Oxyntomodulin
Applications ELISA
Classification Monoclonal
Clone 20
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias GCG, GLP1, GLP2, GRPP
Host Species Mouse
Immunogen Synthetic full length oxyntomodulin coupled to carrier.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.