Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxyntomodulin Antibody (46), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NP261947A40 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP261947A40 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP261947A40 Supplier Novus Biologicals Supplier No. NBP261947AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oxyntomodulin Monoclonal specifically detects Oxyntomodulin in Human samples. It is validated for ELISA.

Specifications

Antigen Oxyntomodulin
Applications ELISA
Classification Monoclonal
Clone 46
Conjugate Alexa Fluor 405
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GCG, GLP1, GLP2, GRPP
Host Species Mouse
Immunogen Synthetic oxyntomodulin 31-37 coupled to carrier
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Thsi antibody is specific for C-terminus of human oxyntomodulin. Does not cross-react with glucagon
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.