Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytocin/Neurophysin I Antibody, Novus Biologicals™
SDP

Catalog No. NBP276346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276346 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276346 Supplier Novus Biologicals Supplier No. NBP276346
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Oxytocin/Neurophysin I Polyclonal specifically detects Oxytocin/Neurophysin I in Mouse, Rat samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.

Specifications

Antigen Oxytocin
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Flow Cytometry, Peptide ELISA Detection limit 1:16000, Immunofluorescence
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias MGC126890, MGC126892, neurophysin I, OT, OT-NPI, oxytocin, prepro- (neurophysin I), oxytocin, prepropeptide, oxytocin-neurophysin 1, oxytocin-neurophysin I, preproprotein
Gene Symbols OXT
Host Species Goat
Immunogen Peptide with sequence DGCRTDPACDPES corresponding to C-Terminus (near) according to Uniprot Mouse NP_035155.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 5020.0
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.