Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxytocin/Neurophysin I Antibody (OTI5G4), PE, Novus Biologicals™
SDP

Catalog No. NB302140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302140 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302140 Supplier Novus Biologicals Supplier No. NBP273178PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oxytocin/Neurophysin I Monoclonal antibody specifically detects Oxytocin/Neurophysin I in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Oxytocin/Neurophysin I
Applications Western Blot
Classification Monoclonal
Clone OTI5G4
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias neurophysin I, OT, OT-NPI, OXT, OXT-NPI, oxytocin, prepro- (neurophysin I), oxytocin, prepropeptide, oxytocin-neurophysin 1, oxytocin-neurophysin I, preproprotein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 20-125 of human Oxytocin/Neurophysin I (NP_000906) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5020
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.