Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ P 22077
GREENER_CHOICE

Catalog No. 448510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-851-0 10 mg
44-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-851-0 Supplier Tocris Bioscience™ Supplier No. 4485/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

USP7 inhibitor

Inhibitor of ubiquitin-specific protease (USP) 7 (EC50 = 8.6 μM); also inhibits the closely related deubiquitinase (DUB) USP47. Demonstrates downstream inhibition of HDM2 and claspin in vitro. Inhibits USP7-mediated p53 deubiquitination. Blocks deubiquitination of Tip60 (KAT5) histone lysine acetyltransferase.

Chemical Identifiers

CAS 1247819-59-5
Molecular Formula C12H7F2NO3S2

Specifications

Quantity 10 mg
Formula Weight 315.32
Purity >99%
Molecular Weight (g/mol) 315.31
Chemical Name or Material P 22077
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.