Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ P 22077
GREENER_CHOICE

Catalog No. p-4816987
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

USP7 inhibitor

Inhibitor of ubiquitin-specific protease (USP) 7 (EC50 = 8.6 μM); also inhibits the closely related deubiquitinase (DUB) USP47. Demonstrates downstream inhibition of HDM2 and claspin in vitro. Inhibits USP7-mediated p53 deubiquitination. Blocks deubiquitination of Tip60 (KAT5) histone lysine acetyltransferase.

Chemical Identifiers

CAS 1247819-59-5
Molecular Formula C12H7F2NO3S2

Specifications

Quantity 10 mg
Formula Weight 315.32
Purity >99%
Molecular Weight (g/mol) 315.31
Chemical Name or Material P 22077
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.