Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Cadherin Antibody (106020) [PerCP], Novus Biologicals™
SDP

Catalog No. NB374357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374357 Supplier Novus Biologicals Supplier No. FAB761C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

P-Cadherin Monoclonal specifically detects P-Cadherin in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen P-Cadherin
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 106020
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse P-Cadherin, Glu100-Gly647, Accession # Q8BSL6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1001
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.