Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Cadherin Antibody (MM0508-9V11), PE, Novus Biologicals™
SDP

Catalog No. NBP211842PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211842PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211842PE Supplier Novus Biologicals Supplier No. NBP211842PE

Mouse Monoclonal Antibody

P-Cadherin Monoclonal specifically detects P-Cadherin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen P-Cadherin
Applications Flow Cytometry
Classification Monoclonal
Clone MM0508-9V11
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. CAA45177
Gene Alias cadherin 3, P-cadherin (placental), cadherin 3, type 1, P-cadherin (placental), cadherin-3, CDHPcalcium-dependent adhesion protein, placental, HJMD, PCADP-cadherin, Placental cadherin
Gene Symbols CDH3
Host Species Mouse
Immunogen Human recombinant P-Cadherin (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1001
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.