Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals™
P-Nitrophenyl-α-D-Galactopyranoside, MP Biomedicals is used as a substrate for α-galactosidase.
$87.00 - $168.00
Specifications
Form | Powder |
---|---|
Format | White Powder |
Molecular Weight (g/mol) | 301.3 |
Storage Requirements | 0°C, protect from light |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP021024712
|
MP Biomedicals™
02102471.2 |
100mg |
Each for $87.00
|
|
|||||
MP021024715
|
MP Biomedicals™
02102471.5 |
500mg |
Each for $168.00
|
|
|||||
Description
Specifications
Powder | |
White Powder | |
301.3 | |
0°C, protect from light |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title