Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p-Phenylenediamine, 99+%
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 139.0°C to 141.0°C |
| Color | Brown to Purple or Gray-White |
| Flash Point | 156°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H4(NH2)2 |
| Quantity | 5 g |
| Beilstein | 13, 61 |
| Merck Index | 15, 7397 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. Toxic in contact with skin. May cause an allergic skin reaction. Toxic if inhaled. Causes serious eye irritation. Toxic if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim |
| EINECSNumber | 203-404-7 |
| RTECSNumber | SS8050000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction