Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech.
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Boiling Point | 148°C to 152°C (1.0 mmHg) |
| Flash Point | >93°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5320 to 1.536 |
| Quantity | 1 g |
| Synonym | methyl 4-1-hydroxyethyl benzoate, 4-1-hydroxyethyl benzoic acid methyl ester, 4-1-hydroxy-ethyl-benzoic acid methyl ester, benzoic acid, 4-1-hydroxyethyl-, methyl ester, acmc-20m5ni, benzoic acid, 4-1s-1-hydroxyethyl-, methyl ester, methyl 4-hydroxyethyl benzoate, methyl4-1-hydroxyethyl benzoate, methyl-4-1-hydroxyethyl benzoate, methyl 4-1-hydroxyethyl benzoate # |
| Solubility Information | Solubility in water: slightly |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction