Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Rex1 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB056267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056267 Supplier Novus Biologicals Supplier No. NBP298883V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P-Rex1 Polyclonal specifically detects P-Rex1 in Human samples. It is validated for Western Blot.

Specifications

Antigen P-Rex1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human P-Rex1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57580
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.