Learn More
Description
Specifications
Specifications
| Antigen | P-Rex1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:750, ELISA 1:20000-1:50000, Immunohistochemistry 1:50-1:150, Immunoprecipitation 1:500, Immunohistochemistry-Paraffin 1:50-1:150 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, ptdIns(34,5)-dependent Rac exchanger 1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to aino acid region 815-836 of Human PREX1. Peptide was covalently modified. Sequence: QEEDQADSAFPLLSLGPRLSLC |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
