Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin/CD62P Antibody (Psel.KO.2.12), PE, Novus Biologicals™
SDP

Catalog No. NB10065391Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065391Q 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065391Q Supplier Novus Biologicals Supplier No. NB10065391PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.

Specifications

Antigen P-Selectin/CD62P
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone Psel.KO.2.12
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. P16109
Gene Alias CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Gene Symbols SELP
Host Species Mouse
Immunogen P-Selectin transfected 300.19 cells.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Test Specificity NB100-65391 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells. Clone Psel.KO.2.12 is reported to inhibit P-selectin-dependent adhesion between HL60 cells and P-selectin transfected COS cells.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.