Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P-Selectin/CD62P Antibody (Psel.KO.2.7), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N1065392PC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1065392PC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1065392PC5 Supplier Novus Biologicals Supplier No. NB10065392PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen P-Selectin/CD62P
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone Psel.KO.2.7
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Gene Symbols SELP
Host Species Mouse
Immunogen P-selectin transfected 300.19 cells.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6403.0
Test Specificity NB100-65392 recognizes the P-Selectin cell surface antigen, a 140kD glycoprotein. P-Selectin is expressed by activated platelets and endothelial cells, and plays an important role in adhesive processes between leucocytes and endothelial cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.