Learn More
RelA/NFkB p65 NLS (p Ser526) Rabbit anti-Human, Clone: SP07-00, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | RelA/NFkB p65 NLS (p Ser526) |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | SP07-00 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10 - 1:500 |
| Formulation | TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide |
| Gene Alias | MGC131774, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65 NLS, p65 Nuclear Localization Sequence, p65-NLS, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 |
| Gene Symbols | RELA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.