Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl chlorothionoformate, 97%

Catalog No. p-7027051
Click to view available options
Quantity:
10 g
50 g

CAS: 937-63-3 | C8H7ClOS | 186.65 g/mol

p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2'-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2′-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.

Solubility
Miscible with common organic solvents.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-63-3
Molecular Formula C8H7ClOS
Molecular Weight (g/mol) 186.65
MDL Number MFCD00014466
InChI Key UNCAXIZUVRKBMN-UHFFFAOYSA-N
Synonym 4-methylphenyl chlorothioformate, o-p-tolyl chlorothioformate, p-tolyl chlorothionoformate, 4-tolyl chlorothionoformate, 4-methylphenyl chloromethanethioate, o-4-methylphenyl chlorothioformate, carbonochloridothioic acid, o-4-methylphenyl ester, o-4-methylphenyl carbonochloridothioate, chlorothioformic acid p-tolyl ester, 4-methylphenoxy methanethioyl chloride
PubChem CID 70305
IUPAC Name O-(4-methylphenyl) chloromethanethioate
SMILES CC1=CC=C(OC(Cl)=S)C=C1

Specifications

Density 1.21
Boiling Point 70°C to 72°C (1 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.574
Quantity 10 g
UN Number UN3265
Beilstein 1448986
Sensitivity Moisture sensitive
Solubility Information Miscible with common organic solvents.
Formula Weight 186.66
Percent Purity 97%
Chemical Name or Material p-Tolyl chlorothionoformate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 213-333-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.