Learn More
p-Xylene, 99%
Description
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
p-Xylene is used as a precursor in the production of benzoic, isophthalic, tetraphillic acids and dimethyle esters, which are used in the manufacture of polyester. It acts as an intermediate in plastic and rubber products.
Solubility
Miscible with alcohol, ether, acetone, benzene and chloroform. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 12°C to 13°C |
| Density | 0.866 |
| Boiling Point | 137°C to 138°C |
| Quantity | 500 mL |
| Flash Point | 27°C (80°F) |
| Odor | Aromatic |
| Refractive Index | 1.495 |
| UN Number | UN1307 |
| Beilstein | 1901563 |
| Merck Index | 14,10081 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H312-H332-H315 Flammable liquid and vapor. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES |
| EINECSNumber | 203-396-5 |
| RTECSNumber | ZE2625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.