Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Antibody (25a), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP24411A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP24411A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP24411A70 Supplier Novus Biologicals Supplier No. NBP254411AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p120-catenin Monoclonal specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p120-catenin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 25a
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Gene Symbols CTNND1
Host Species Mouse
Immunogen Mouse p120-catenin (pY96) (Uniprot: O60716)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.