Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Antibody (CTNND1/4383R), FITC, Novus Biologicals™
SDP

Catalog No. NB385535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385535 Supplier Novus Biologicals Supplier No. NBP308618F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

p120-catenin Monoclonal antibody specifically detects p120-catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen p120-catenin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CTNND1/4383R
Conjugate FITC
Formulation PBS
Gene Alias Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Host Species Rabbit
Immunogen Recombinant fragment (around aa900-1,000) of human p120-catenin protein (exact sequence is proprietary) (Uniprot: O60716)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.