Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p120-catenin Antibody (OTI2E8), FITC, Novus Biologicals™
SDP

Catalog No. NBP273179F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273179F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273179F Supplier Novus Biologicals Supplier No. NBP273179F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p120-catenin Monoclonal specifically detects p120-catenin in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry.

Specifications

Antigen p120-catenin
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2E8
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN
Gene Symbols CTNND1
Host Species Mouse
Immunogen Full length recombinant protein of human p120-catenin (NP_001078927) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1500.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.