Learn More
p130Cas Ab-1 Mouse Monoclonal Antibody, Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ p130Cas Ab-1, Mouse Monoclonal Antibody.
Specifications
Antigen | p130superscriptCassuperscript Ab-1 |
---|---|
Clone | CAS-14; same as 35B-1A4 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
MS855P0
|
Epredia
MS855P0 |
Concentrated | 200μg/mL | 100 μL | N/A | N/A | N/A | ||
MS855P1
|
Epredia
MS855P1 |
Concentrated | 200μg/mL | 500 μL | N/A | N/A | N/A | ||
MS855P
|
Epredia
MS855P |
Concentrated | 200μg/mL | 1 mL | N/A | N/A | N/A | ||
MS855P1ABX
|
Epredia
MS855P1ABX |
Concentrated | 1mg/mL | 100 μL | N/A | N/A | N/A | ||
MS855PABX
|
Epredia
MS855PABX |
Concentrated | 1mg/mL | 200 μL | N/A | N/A | N/A | ||
MS855R1
|
Epredia
MS855R1 |
Prediluted | — | 1 mL | N/A | N/A | N/A | ||
MS855R7
|
Epredia
MS855R7 |
Prediluted | — | 7 mL | N/A | N/A | N/A | ||
Description
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich region p130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.Host Species: Mouse
Clone: CAS-14; same as 35B-1A4
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant human p130Cas protein
Molecular Weight: 130kDa
Positive Control: MCF-7 Cells, HUVEC cells, Tonsil or breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
p130superscriptCassuperscript Ab-1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Mouse | |
Cell Cycle | |
Recombinant human p130superscriptCassuperscript protein | |
Primary |
CAS-14; same as 35B-1A4 | |
Monoclonal | |
RUO | |
Human, Rat | |
IgG2a κ | |
MCF-7 cells, HUVEC cells, tonsil or breast CA |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.