Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p130Cas/BCAR1 Antibody (OTI1A9), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273181C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273181C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273181C Supplier Novus Biologicals Supplier No. NBP273181C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p130Cas/BCAR1 Monoclonal specifically detects p130Cas/BCAR1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen p130Cas/BCAR1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A9
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas
Gene Symbols BCAR1
Host Species Mouse
Immunogen Full length recombinant protein of human p130Cas/BCAR1 (NP_055382) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9564.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.