Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p14ARF/CDKN2A Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N200111J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N200111J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N200111J646 Supplier Novus Biologicals Supplier No. NB200111JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p14ARF/CDKN2A Polyclonal specifically detects p14ARF/CDKN2A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen p14ARF/CDKN2A
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, P16, p16-INK4, p16-INK4a, P16INK4A, P19, p19Arf, TP16
Gene Symbols CDKN2A
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of human p14ARF (between residues 50-150). [Swiss-Prot No. Q8N726]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1029.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.