Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p15INK4b/CDKN2B Antibody (OTI3B6), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N273184P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273184P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273184P594 Supplier Novus Biologicals Supplier No. NBP273184PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

p15INK4b/CDKN2B Monoclonal specifically detects p15INK4b/CDKN2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen p15INK4b/CDKN2B
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B6
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CDK4I, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15
Gene Symbols CDKN2B
Host Species Mouse
Immunogen Full length recombinant protein of human p15INK4b/CDKN2B (NP_004927) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1030.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.