Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P15RS Antibody, Novus Biologicals™
SDP

Catalog No. NBP219650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219650 Supplier Novus Biologicals Supplier No. NBP219650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

P15RS Polyclonal specifically detects P15RS in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen P15RS
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.69mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96P16
Gene Alias Cyclin-dependent kinase inhibitor 2B-related protein, Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein), FLJ10656, HsT3101, MGC19513, p15INK4B-related protein, P15RScyclin-dependent kinase 2B-inhibitor-related protein, regulation of nuclear pre-mRNA domain containing 1A, regulation of nuclear pre-mRNA domain-containing protein 1A
Gene Symbols RPRD1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human P15RS. The exact sequence is proprietary.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 55197
Test Specificity Human. Species reactivity based on sequence identity: Chimpanzee (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.