Learn More
P16-INK4A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | P16-INK4A |
---|---|
Concentration | 0.37 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-553-876
![]() ![]() |
Proteintech Group Inc
108831AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
10883-1-AP reacts with p16 specifically. P16 plays an essential role in regulating the cell cycle, and mutations in p16 increase the risk of developing various cancers, including melanoma.
Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.Specifications
P16-INK4A | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q8N726 | |
1029 | |
P16-INK4A Fusion Protein Ag1328 | |
Primary | |
-20°C | |
CDKN2A |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARF, ARF,CDKN2A, CDK4 inhibitor p16 INK4, CDK4 inhibitor p16INK4, CDK4I, CDKN2, CDKN2A, CMM2, INK4, INK4a, MLM, MTS 1, MTS1, Multiple tumor suppressor 1, P16, p16 INK4, p16 INK4a, p16INK4, p16INK4a, TP16 | |
CDKN2A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.