Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p16INK4a/CDKN2A Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB180266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180266 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180266 Supplier Novus Biologicals Supplier No. NBP306986C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

p16INK4a/CDKN2A Polyclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen p16INK4a/CDKN2A
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human CDKN2A. The immunogen is located within the last 50 amino acids of CDKN2A.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1029
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.