Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p18INK4c/CDKN2C Antibody (OTI2E1), PerCP, Novus Biologicals™
SDP

Catalog No. NB302134 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302134 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302134 Supplier Novus Biologicals Supplier No. NBP273185PCP

Mouse Monoclonal Antibody

p18INK4c/CDKN2C Monoclonal antibody specifically detects p18INK4c/CDKN2C in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen p18INK4c/CDKN2C
Applications Western Blot
Classification Monoclonal
Clone OTI2E1
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias CDK6 inhibitor p18, CDKN6, cyclin-dependent inhibitor, cyclin-dependent kinase 4 inhibitor C, Cyclin-dependent kinase 6 inhibitor, cyclin-dependent kinase inhibitor 2C (p18, inhibits CDK4), INK4Ccyclin-dependent kinase 6 inhibitor p18, p18, p18-INK4C, p18-INK6
Host Species Mouse
Immunogen Full length human recombinant protein of human CDKN2C (NP_001253) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1031
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.