Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19 INK4d Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB322805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322805 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322805 Supplier Novus Biologicals Supplier No. NBP300015JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p19 INK4d Polyclonal antibody specifically detects p19 INK4d in Human samples. It is validated for ELISA

Specifications

Antigen p19 INK4d
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human p19 INK4d (Accession#: P55273; Met10-Leu166)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1032
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.