Learn More
Description
Specifications
Specifications
| Antigen | p19 INK4d |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | DCS100 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Formulation | PBS |
| Gene Alias | CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D |
| Host Species | Mouse |
| Immunogen | A his-tagged recombinant full length p19 protein recognizing a 19kD protein (Uniprot: P55273) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
