Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190RhoGAP Antibody, Novus Biologicals™
SDP

Catalog No. NB10088154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10088154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10088154 Supplier Novus Biologicals Supplier No. NB10088154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

p190RhoGAP Polyclonal specifically detects p190RhoGAP in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen p190RhoGAP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9NRY4
Gene Alias glucocorticoid receptor DNA binding factor 1, glucocorticoid receptor DNA-binding factor 1, glucocorticoid receptor repression factor 1, GRF1, GRF-1, GRLF1, KIAA1722, MGC10745, P190A, P190-A, p190ARhoGAP, p190RhoGAP, rho GAP p190A, Rho GTPase activating protein 35, rho GTPase-activating protein 35
Gene Symbols ARHGAP35
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1200 and 1250 of human glucocorticoid receptor DNA binding factor 1 (p190RHOGAP) using the numbering given in entry EAW57452.1 (GeneID 2909).
Molecular Weight of Antigen 172 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2909
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, Crabeating macaque, European domestic ferret, African elephant and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.