Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p19ARF/CDKN2A Antibody (5-C3-1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013970 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013970 Supplier Novus Biologicals Supplier No. NBP280897
Add to Cart
Add to Cart

Rat Monoclonal Antibody

p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready, Knockout Validated.

Specifications

Antigen p19ARF/CDKN2A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF, Immunoassay
Classification Monoclonal
Clone 5-C3-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500 - 1:1000, Flow Cytometry : 1 ug/ml, Immunohistochemistry : 5 ug/ml, Immunocytochemistry/Immunofluorescence : 1:500 - 1:1000, Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen : 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated
Formulation PBS
Gene Alias alternative reading frame, ARF, ARF-INK4a, CDK4I, CDKN2A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4), cyclin-dependent kinase inhibitor 2A, isoform 3, cyclin-dependent kinase inhibitor 2A, isoforms 1/2, cyclin-dependent kinase inhibitor protein, INK4, INK4a, Ink4a/Arf, INK4a-ARF, mitochondrial smARF', MTS1, p14, p14ARF, P16, p16(INK4a), p16-INK4, p16INK4a, p16-INK4a, P19, p19<ARF>, p19ARF, Pctr1
Gene Symbols Cdkn2a
Host Species Rat
Immunogen A synthetic peptide containing amino acids 54-75 of the murine p19ARF/CDKN2A. [UniProt# Q64364]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 12578
Test Specificity Human reactivity reported in scientific literature (PMID: 27177758). The antibody does not react with Golden Syrian hamster. Armenian hamster has not been tested.
Target Species Human, Mouse, Hamster (Negative)
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.